4-Chloro-3-(trifluoromethyl)benzyl alcohol ≥95%

Supplier: Matrix Scientific

Synonyms: (4-Chloro-3-(trifluoromethyl)phenyl)methanol

002307-1G 002307-5G
101783-658EA 36.92 USD
101783-658 101783-660
4-Chloro-3-(trifluoromethyl)benzyl alcohol ≥95%
4-Chloro-3-(trifluoromethyl)benzyl alcohol

Hazard: Irritant

Formula: C₈H₆ClF₃O
MW: 210.5809 g/mol
MDL Number: MFCD01631538
CAS Number: 65735-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR