2-Fluoro-3-(trifluoromethyl)cinnamic acid

Supplier: Matrix Scientific

002300-1G 002300-5G
101783-638EA 36.92 USD
101783-638 101783-640
2-Fluoro-3-(trifluoromethyl)cinnamic acid
2-Fluoro-3-(trifluoromethyl)cinnamic acid

Hazard: Irritant

Formula: C₁₀H₆F₄O₂
MDL Number: MFCD00236291
CAS Number: 237069-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR