4-Fluoro-3-(trifluoromethyl)phenylacetic acid ≥98%

Supplier: Matrix Scientific

002292-5G 002292-25G
101783-622EA 27.69 USD
101783-622 101783-624
4-Fluoro-3-(trifluoromethyl)phenylacetic acid ≥98%
4-Fluoro-3-(trifluoromethyl)phenylacetic acid

Hazard: Irritant

Formula: C₉H₆F₄O₂
MW: 222.14 g/mol
MDL Number: MFCD00061190
CAS Number: 220227-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR