4-Fluoro-3-(trifluoromethyl)benzophenone ≥97%
Supplier: Matrix Scientific
002283-1G
002283-5G
101783-606EA
44.62
USD
101783-606
101783-608
4-Fluoro-3-(trifluoromethyl)benzophenone ≥97%
4-Fluoro-3-(trifluoromethyl)benzophenone
Hazard: Irritant
Formula:
C₁₄H₈F₄O MW: 268.21 g/mol |
MDL Number:
MFCD00061258 CAS Number: 239087-04-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...