2-Fluoro-3-(trifluoromethyl)benzophenone ≥97%

Supplier: Matrix Scientific

002282-1G 002282-5G
101783-602EA 44.62 USD
101783-602 101783-604
2-Fluoro-3-(trifluoromethyl)benzophenone ≥97%
2-Fluoro-3-(trifluoromethyl)benzophenone

Hazard: Irritant

Formula: C₁₄H₈F₄O
MDL Number: MFCD00061151
CAS Number: 207853-70-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR