2-Fluoro-3-(trifluoromethyl)benzamide ≥97%
Supplier: Matrix Scientific
002279-1G
002279-5G
101783-590EA
44.62
USD
101783-590
101783-592
2-Fluoro-3-(trifluoromethyl)benzamide ≥97%
2-Fluoro-3-(trifluoromethyl)benzamide
Hazard: Irritant
Formula:
C₈H₅F₄NO |
MDL Number:
MFCD00061146 CAS Number: 207853-60-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...