4-Chloro-3-(trifluoromethyl)-5-(phenyl)pyrazole ≥97%

Supplier: Matrix Scientific

002277-1G 002277-5G
101783-582EA 35.38 USD
101783-582 101783-584
4-Chloro-3-(trifluoromethyl)-5-(phenyl)pyrazole ≥97%
4-Chloro-3-(trifluoromethyl)-5-(phenyl)pyrazole

Hazard: Irritant

Formula: C₁₀H₆ClF₃N₂
MDL Number: MFCD00155725
CAS Number: 235106-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR