2-Chloro-3,6-difluoroanisole

Supplier: Matrix Scientific

002260-1G 002260-5G
101783-556EA 63.08 USD
101783-556 101783-558
2-Chloro-3,6-difluoroanisole
2-Chloro-3,6-difluoroanisole

Hazard: Irritant

Formula: C₇H₅ClF₂O
MDL Number: MFCD01631451
CAS Number: 261762-37-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR