5-Chloro-1,3-benzodioxole

Supplier: Matrix Scientific

Synonyms: 5-Chlorobenzo[d][1,3]dioxole

002254-1G 002254-5G
101783-534EA 38.46 USD
101783-534 101783-536
5-Chloro-1,3-benzodioxole
5-Chloro-1,3-benzodioxole

Hazard: Irritant

Formula: C₇H₅ClO₂
MW: 156.57 g/mol
Boiling Pt: 65-69°C
MDL Number: MFCD00010842
CAS Number: 7228-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR