2-Chloro-3,4,4,4-tetrafluoro-3-(trifluoromethyl)-1-butene ≥97%

Supplier: Matrix Scientific

002252-5G 002252-25G
101783-530EA 141.54 USD
101783-530 101783-532
2-Chloro-3,4,4,4-tetrafluoro-3-(trifluoromethyl)-1-butene ≥97%
2-Chloro-3,4,4,4-tetrafluoro-3-(trifluoromethyl)-1-butene

Hazard: Irritant

Formula: C₅H₂ClF₇
MW: 230.51 g/mol
MDL Number: MFCD00155721
CAS Number: 235106-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR