(5-Fluoro-2-methylphenyl)acetic acid ≥97%
Supplier: Matrix Scientific
002183-1G
002183-5G
002183-25G
101783-412EA
76.92
USD
101783-412
101783-414
101783-416
(5-Fluoro-2-methylphenyl)acetic acid ≥97%
(5-Fluoro-2-methylphenyl)acetic acid
Hazard: Irritant
Formula:
C₉H₉FO₂ MW: 168.17 |
MDL Number:
MFCD01631530 CAS Number: 261951-75-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...