4-Chloro-2-methylbenzylamine
Supplier: Matrix Scientific
002172-1G
002172-5G
101783-382EA
78.46
USD
101783-382
101783-384
4-Chloro-2-methylbenzylamine
4-Chloro-2-methylbenzylamine
Hazard: Irritant
Formula:
C₈H₁₀ClN |
MDL Number:
MFCD00047932 CAS Number: 27917-11-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...