5-Chloro-2-methoxyphenylboronic acid ≥97%
Supplier: Matrix Scientific
002150-5G
002150-25G
101783-338EA
40
USD
101783-338
101783-340
5-Chloro-2-methoxyphenylboronic acid ≥97%
5-Chloro-2-methoxyphenylboronic acid
Hazard: Irritant/Store in Freezer
Formula:
C₇H₈BClO₃ Melting Pt: 141…142 °C |
MDL Number:
MFCD01318966 CAS Number: 89694-48-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...