5-Chloro-2-fluorophenylacetonitrile ≥97%

Supplier: Matrix Scientific

002115-5G 002115-25G
101783-236EA 27.69 USD
101783-236 101783-238
5-Chloro-2-fluorophenylacetonitrile ≥97%
5-Chloro-2-fluorophenylacetonitrile

Hazard: Irritant-Harmful

Formula: C₈H₅ClFN
MDL Number: MFCD01631438
CAS Number: 75279-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR