(5-Chloro-2-fluorophenyl)acetic acid

Supplier: Matrix Scientific

Synonyms: 2-(5-Chloro-2-fluorophenyl)acetic acid

002113-1G 002113-5G
101783-232EA 76.92 USD
101783-232 101783-234
(5-Chloro-2-fluorophenyl)acetic acid
(5-Chloro-2-fluorophenyl)acetic acid

Hazard: Irritant

Formula: (C₆H₃ClF)CH₂COOH
MDL Number: MFCD01631439
CAS Number: 261762-97-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR