3-Chloro-2-fluorocinnamic acid
Supplier: Matrix Scientific
Synonyms:
3-(3-Chloro-2-fluorophenyl)acrylic acid
002105-1G
002105-5G
101783-206EA
67.69
USD
101783-206
101783-208
3-Chloro-2-fluorocinnamic acid
3-Chloro-2-fluorocinnamic acid
Hazard: Irritant-Harmful
Formula:
C₉H₆ClFO₂ MW: 200.6 g/mol |
MDL Number:
MFCD01631392 CAS Number: 261762-62-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...