3-Chloro-2-fluoroanisole

Supplier: Matrix Scientific

Synonyms: 1-Chloro-2-fluoro-3-methoxybenzene

002085-1G 002085-5G
101783-168EA 52.31 USD
101783-168 101783-170
3-Chloro-2-fluoroanisole
3-Chloro-2-fluoroanisole

Hazard: Flammable

Formula: C₇H₆ClFO
MW: 160.58 g/mol
MDL Number: MFCD01631570
CAS Number: 261762-56-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR