3-Chloro-2-fluoro-6-(trifluoromethyl)benzamide

Supplier: Matrix Scientific

002073-1G 002073-5G
101783-156EA 70.77 USD
101783-156 101783-158
3-Chloro-2-fluoro-6-(trifluoromethyl)benzamide
3-Chloro-2-fluoro-6-(trifluoromethyl)benzamide

Hazard: Irritant

Formula: C₈H₄ClF₄NO
MDL Number: MFCD01631400
CAS Number: 186517-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR