3-Chloro-2-fluoro-5-(trifluoromethyl)phenylacetonitrile

Supplier: Matrix Scientific

002071-1G 002071-5G
101783-148EA 109.23 USD
101783-148 101783-150
3-Chloro-2-fluoro-5-(trifluoromethyl)phenylacetonitrile
3-Chloro-2-fluoro-5-(trifluoromethyl)phenylacetonitrile

Hazard: Irritant-Harmful

Formula: C₉H₄ClF₄N
MDL Number: MFCD01631515
CAS Number: 261763-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR