(3-Chloro-2-fluoro-5-(trifluoromethyl)phenyl)acetic acid

Supplier: Matrix Scientific

002070-1G 002070-5G
101783-144EA 173.85 USD
101783-144 101783-146
(3-Chloro-2-fluoro-5-(trifluoromethyl)phenyl)acetic acid
(3-Chloro-2-fluoro-5-(trifluoromethyl)phenyl)acetic acid

Hazard: Irritant

Formula: C₉H₅ClF₄O₂
MDL Number: MFCD01631516
CAS Number: 261763-13-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR