3-Chloro-2-fluoro-5-(trifluoromethyl)benzyl alcohol

Supplier: Matrix Scientific

002065-1G 002065-5G
101783-140EA 44.62 USD
101783-140 101783-142
3-Chloro-2-fluoro-5-(trifluoromethyl)benzyl alcohol
3-Chloro-2-fluoro-5-(trifluoromethyl)benzyl alcohol

Hazard: Irritant

Formula: C₈H₅ClF₄O
MDL Number: MFCD01631512
CAS Number: 261763-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR