3-Chloro-2-fluoro-5-(trifluoromethyl)benzaldehyde

Supplier: Matrix Scientific

002061-1G 002061-5G
101783-128EA 38.46 USD
101783-128 101783-130
3-Chloro-2-fluoro-5-(trifluoromethyl)benzaldehyde
3-Chloro-2-fluoro-5-(trifluoromethyl)benzaldehyde

Hazard: Irritant

Formula: C₈H₃ClF₄O
MW: 226.56 g/mol
MDL Number: MFCD01631510
CAS Number: 261763-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR