(6-Chloro-2-fluoro-3-methylphenyl)acetic acid
Supplier: Matrix Scientific
002057-1G
002057-5G
101783-120EA
169.23
USD
101783-120
101783-122
(6-Chloro-2-fluoro-3-methylphenyl)acetic acid
(6-Chloro-2-fluoro-3-methylphenyl)acetic acid
Hazard: Irritant
Formula:
C₉H₈ClFO₂ MW: 202.61 |
MDL Number:
MFCD01631375 CAS Number: 261762-93-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...