6-Chloro-2-fluoro-3-methylbenzyl alcohol

Supplier: Matrix Scientific

Synonyms: (6-Chloro-2-fluoro-3-methylphenyl)methanol

002053-1G 002053-5G
101783-116EA 47.69 USD
101783-116 101783-118
6-Chloro-2-fluoro-3-methylbenzyl alcohol
6-Chloro-2-fluoro-3-methylbenzyl alcohol

Hazard: Irritant

Formula: C₈H₈ClFO
MW: 174.6 g/mol
Melting Pt: 56…58 °C
MDL Number: MFCD01631372
CAS Number: 261762-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR