6-Chloro-2-fluoro-3-methylbenzamide

Supplier: Matrix Scientific

002050-1G 002050-5G
101783-108EA 101.54 USD
101783-108 101783-110
6-Chloro-2-fluoro-3-methylbenzamide
6-Chloro-2-fluoro-3-methylbenzamide

Hazard: Irritant

Formula: C₈H₇ClFNO
Melting Pt: 150…152 °C
MDL Number: MFCD01631462
CAS Number: 286474-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR