6-Chloro-2-fluoro-3-methylanisole
Supplier: Matrix Scientific
Synonyms:
1-Chloro-3-fluoro-2-methoxy-4-methylbenzene
002048-1G
002048-5G
101783-104EA
61.54
USD
101783-104
101783-106
6-Chloro-2-fluoro-3-methylanisole
6-Chloro-2-fluoro-3-methylanisole
Hazard: Irritant
Formula:
C₈H₈ClFO MW: 174.60 g/mol |
MDL Number:
MFCD01631377 CAS Number: 261762-80-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...