6'-Chloro-2'-fluoro-3'-methylacetophenone
Supplier: Matrix Scientific
002047-1G
002047-5G
101783-100EA
36.92
USD
101783-100
101783-102
6'-Chloro-2'-fluoro-3'-methylacetophenone
6'-Chloro-2'-fluoro-3'-methylacetophenone
Hazard: Irritant
Formula:
C₉H₈ClFO |
MDL Number:
MFCD01631378 CAS Number: 261762-78-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...