5-Chloro-2-(trifluoromethyl)benzyl alcohol

Supplier: Matrix Scientific

Synonyms: (5-Chloro-2-(trifluoromethyl)phenyl)methanol

002004-1G 002004-5G
101783-044EA 158.46 USD
101783-044 101783-046
5-Chloro-2-(trifluoromethyl)benzyl alcohol
5-Chloro-2-(trifluoromethyl)benzyl alcohol

Hazard: Irritant

Formula: C₈H₆ClF₃O
MW: 210.5800 g/mol
MDL Number: MFCD01631355
CAS Number: 261763-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR