5-Chloro-2-(trifluoromethyl)benzoyl chloride

Supplier: Matrix Scientific

002003-1G 002003-5G
101783-040EA 72.31 USD
101783-040 101783-042
5-Chloro-2-(trifluoromethyl)benzoyl chloride
5-Chloro-2-(trifluoromethyl)benzoyl chloride

Hazard: Corrosive

Formula: C₈H₃Cl₂F₃O
Boiling Pt: 214-217°C
MDL Number: MFCD01631354
CAS Number: 320-84-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR