5-Chloro-2-(trifluoromethyl)benzamide
Supplier: Matrix Scientific
002001-1G
002001-5G
101783-032EA
60
USD
101783-032
101783-034
5-Chloro-2-(trifluoromethyl)benzamide
5-Chloro-2-(trifluoromethyl)benzamide
Hazard: Irritant
Formula:
C₈H₅ClF₃NO Melting Pt: 137 °C |
MDL Number:
MFCD01631406 CAS Number: 654-94-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...