5-Fluoro-2-(trifluoromethyl)cinnamic acid

Supplier: Matrix Scientific

001998-1G 001998-5G
101783-024EA 47.69 USD
101783-024 101783-026
5-Fluoro-2-(trifluoromethyl)cinnamic acid
5-Fluoro-2-(trifluoromethyl)cinnamic acid

Hazard: Irritant

Formula: C₁₀H₆F₄O₂
MDL Number: MFCD00236299
CAS Number: 231291-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR