5-Fluoro-2-(trifluoromethyl)benzamide ≥97%

Supplier: Matrix Scientific

001989-1G 001989-5G
101783-008EA 47.69 USD
101783-008 101783-010
5-Fluoro-2-(trifluoromethyl)benzamide ≥97%
5-Fluoro-2-(trifluoromethyl)benzamide

Hazard: Irritant

Formula: C₈H₅F₄NO
Melting Pt: 118…119 °C
CAS Number: 654-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR