4-Bromo-2-chloro-6-fluoroanisole

Supplier: Matrix Scientific

001625-1G 001625-5G
101782-118EA 67.69 USD
101782-118 101782-120
4-Bromo-2-chloro-6-fluoroanisole
4-Bromo-2-chloro-6-fluoroanisole

Hazard: Irritant

Formula: C₇H₅BrClFO
MW: 239.47 g/mol
MDL Number: MFCD01631477
CAS Number: 261762-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR