4-Bromo-2-chloro-5-fluorobenzoic acid ≥97%

Supplier: Matrix Scientific

001617-1G 001617-5G 001617-25G
101782-098EA 21.54 USD
101782-098 101782-100 102663-540
4-Bromo-2-chloro-5-fluorobenzoic acid ≥97%
4-Bromo-2-chloro-5-fluorobenzoic acid

Hazard: Irritant

Formula: C₇H₃BrClFO₂
MDL Number: MFCD00236589
CAS Number: 177480-81-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR