4-Hydroxy-6-(trifluoromethyl)quinoline-3-carboxylic acid ≥97%
Supplier: Matrix Scientific
001514-1G
001514-5G
101781-862EA
112.31
USD
101781-862
101781-864
4-Hydroxy-6-(trifluoromethyl)quinoline-3-carboxylic acid ≥97%
4-Hydroxy-6-(trifluoromethyl)quinoline-3-carboxylic acid
Hazard: Irritant
Formula:
C₁₁H₆F₃NO₃ |
MDL Number:
MFCD00236691 CAS Number: 49713-47-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...