3'-Fluoro-5'-(trifluoromethyl)acetophenone ≥98%

Supplier: Matrix Scientific

Synonyms: 1-[3-Fluoro-5-(trifluoromethyl)phenyl]ethan-1-one, 3-Acetyl-5-fluorobenzotrifluoride

001497-1G 001497-5G
101781-822EA 44.62 USD
101781-822 101781-824
3'-Fluoro-5'-(trifluoromethyl)acetophenone ≥98%
3'-Fluoro-5'-(trifluoromethyl)acetophenone

Hazard: Irritant

Formula: C₉H₆F₄O
MW: 206.1370 g/mol
MDL Number: MFCD00061250
CAS Number: 202664-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR