3'-Fluoro-4'-(trifluoromethyl)acetophenone ≥98%

Supplier: Matrix Scientific

Synonyms: 1-(3-Fluoro-4-(trifluoromethyl)phenyl)ethanone

001473-1G 001473-5G
101781-754EA 92.31 USD
101781-754 101781-756
3'-Fluoro-4'-(trifluoromethyl)acetophenone ≥98%
3'-Fluoro-4'-(trifluoromethyl)acetophenone

Hazard: Irritant

Formula: C₉H₆F₄O
MW: 206.14 g/mol
MDL Number: MFCD00236273
CAS Number: 237761-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR