4-Methoxy-3-(trifluoromethyl)benzamide

Supplier: Matrix Scientific

001445-1G 001445-5G
101781-674EA 41.54 USD
101781-674 101781-676
4-Methoxy-3-(trifluoromethyl)benzamide
4-Methoxy-3-(trifluoromethyl)benzamide

Hazard: Irritant

Formula: C₉H₈F₃NO₂
MDL Number: MFCD01631493
CAS Number: 261951-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR