4'-Fluoro-3'-(trifluoromethyl)acetophenone ≥98%

Supplier: Matrix Scientific

001438-5G 001438-25G 001438-100G
101781-648EA 29.23 USD
101781-648 101781-650 101781-652
4'-Fluoro-3'-(trifluoromethyl)acetophenone ≥98%
4'-Fluoro-3'-(trifluoromethyl)acetophenone

Hazard: Irritant

Formula: C₉H₆F₄O
MDL Number: MFCD00061252
CAS Number: 208173-24-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR