2-(Methylsulfanyl)-6-(trifluoromethyl)-4-pyrimidinol ≥97%

Supplier: Matrix Scientific

Synonyms: 2-(Methylthio)-6-(trifluoromethyl)pyrimidin-4-ol, 4-Hydroxy-2-(methylthio)-6-(trifluoromethyl)pyrimidine, 2-(Methylthio)-6-(trifluoromethyl)pyrimidin-4(1H)-one

001422-1G 001422-5G
101781-624EA 29.23 USD
101781-624 101781-626
2-(Methylsulfanyl)-6-(trifluoromethyl)-4-pyrimidinol ≥97%
2-(Methylsulfanyl)-6-(trifluoromethyl)-4-pyrimidinol

Hazard: Irritant

Formula: C₆H₅F₃N₂OS
MW: 210.18 g/mol
Melting Pt: 181…182 °C
MDL Number: MFCD00153191
CAS Number: 16097-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR