4'-Chloro-2'-fluoro-5'-methylacetophenone

Supplier: Matrix Scientific

Synonyms: 1-(4-Chloro-2-fluoro-5-methylphenyl)ethanone

001406-250MG 001406-1G
101781-580EA 55.38 USD
101781-580 101781-582
4'-Chloro-2'-fluoro-5'-methylacetophenone
4'-Chloro-2'-fluoro-5'-methylacetophenone

Hazard: Irritant

Formula: C₉H₈ClFO
MW: 186.6106 g/mol
MDL Number: MFCD00236601
CAS Number: 177211-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR