2-Amino-6-(trifluoromethylthio)benzothiazole

Supplier: Matrix Scientific

Synonyms: 6-((Trifluoromethyl)thio)benzo[d]thiazol-2-amine

001363-250MG 001363-1G
101781-496EA 192.31 USD
101781-496 101781-498
2-Amino-6-(trifluoromethylthio)benzothiazole
2-Amino-6-(trifluoromethylthio)benzothiazole

Hazard: Irritant

Formula: C₈H₅F₃N₂S₂
MW: 250.27 g/mol
Melting Pt: 153…154 °C
MDL Number: MFCD00665748
CAS Number: 326-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR