4-Amino-8-(trifluoromethyl)quinoline

Supplier: Matrix Scientific

Synonyms: 8-(Trifluoromethyl)quinolin-4-amine

001361-250MG 001361-1G
101781-488EA 153.85 USD
101781-488 101781-490
4-Amino-8-(trifluoromethyl)quinoline
4-Amino-8-(trifluoromethyl)quinoline

Hazard: Irritant

Formula: C₁₀H₇F₃N₂
MW: 212.18 g/mol
MDL Number: MFCD00269346
CAS Number: 243977-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR