4-Amino-7-(trifluoromethyl)quinoline

Supplier: Matrix Scientific

001360-250MG 001360-1G
101781-484EA 203.08 USD
101781-484 101781-486
4-Amino-7-(trifluoromethyl)quinoline
4-Amino-7-(trifluoromethyl)quinoline

Hazard: Irritant

Formula: C₁₀H₇F₃N₂
MW: 212.18 g/mol
MDL Number: MFCD00269411
CAS Number: 243666-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR