4-Amino-6-(trifluoromethyl)quinoline

Supplier: Matrix Scientific

Synonyms: 6-(Trifluoromethyl)quinolin-4-amine

001359-250MG 001359-1G
101781-480EA 140 USD
101781-480 101781-482
4-Amino-6-(trifluoromethyl)quinoline
4-Amino-6-(trifluoromethyl)quinoline

Hazard: Irritant

Formula: C₁₀H₇F₃N₂
MW: 212.17 g/mol
MDL Number: MFCD00278781
CAS Number: 247113-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR