2,2,3,3,4,4,5,5-Octafluoro-1,6-hexyldimethacrylate ≥97%

Supplier: Matrix Scientific

001220-1G 001220-5G
101781-190EA 80 USD
101781-190 101781-192
2,2,3,3,4,4,5,5-Octafluoro-1,6-hexyldimethacrylate ≥97%
2,2,3,3,4,4,5,5-Octafluoro-1,6-hexyldimethacrylate

Hazard: Irritant/Refrigerate

Formula: C₁₄H₁₄F₈O₄
MW: 398.25 g/mol
MDL Number: MFCD00078359
CAS Number: 66818-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR