1,1,2,3,3,3-Hexafluoropropyl-2,2,2-trifluoroethyl ether ≥97%

Supplier: Matrix Scientific

001185-5G 001185-25G
101781-110EA 272.31 USD
101781-110 101781-112
1,1,2,3,3,3-Hexafluoropropyl-2,2,2-trifluoroethyl ether ≥97%
1,1,2,3,3,3-Hexafluoropropyl-2,2,2-trifluoroethyl ether

Hazard: Irritant

Formula: C₅H₃F₉O
MW: 250.06 g/mol
Boiling Pt: 72 °C (750 Torr)
Density: 1.5398
MDL Number: MFCD00476468
CAS Number: 993-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR