4-Allyl-2,3,5,6-tetrafluorobenzoic acid ≥97%

Supplier: Matrix Scientific

001179-1G 001179-5G
101781-098EA 200 USD
101781-098 101781-100
4-Allyl-2,3,5,6-tetrafluorobenzoic acid ≥97%
4-Allyl-2,3,5,6-tetrafluorobenzoic acid

Hazard: Irritant

Formula: C₁₀H₆F₄O₂
Melting Pt: 89…90 °C
MDL Number: MFCD00082610
CAS Number: 79538-02-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR