2,2-Dimethyl-3-hexanone ≥98%
Supplier: Matrix Scientific
Synonyms:
2,2-Dimethylhexan-3-one
032354-1G
032354-5G
032354-25G
101833-550EA
44.62
USD
101833-550
101833-552
101833-554
2,2-Dimethyl-3-hexanone ≥98%
2,2-Dimethyl-3-hexanone
Hazard: Irritant
Formula:
C₈H₁₆O |
MDL Number:
MFCD00027140 CAS Number: 5405-79-8 |
Specification Test Results
ND20 | 1.4105 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...