2,3-Dibromopropyl isothiocyanate

Supplier: Matrix Scientific

004589-5G 004589-25G
101787-538EA 80 USD
101787-538 101787-540
2,3-Dibromopropyl isothiocyanate
2,3-Dibromopropyl isothiocyanate

Hazard: Toxic/Refrigerate/Lachrymator

Formula: C₄H₅Br₂NS
Density: 1.96
MDL Number: MFCD00060411
CAS Number: 51784-10-2

Order Now

Specification Test Results

Density 1.96

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR